Key Documents
CLS3883
Corning® 96 Well Half-Area Microplate
flat bottom clear, white polystyrene, bag of 25, non-sterile, lid: no
Sinonimo/i:
96 multiwell plate
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.15
Materiali
flat bottom clear
white polystyrene
Descrizione
non-treated
Sterilità
non-sterile
Caratteristiche
lid: no
Confezionamento
bag of 25
pkg of 25 inner package
Produttore/marchio commerciale
Corning 3883
Prof. pozzetto
10.7 mm
Descrizione generale
Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.
Note legali
Corning is a registered trademark of Corning, Inc.