CLS2980225
Corning® cover glasses
rectangular, No. 11/2, W × L 22 mm × 50 mm
Sinonimo/i:
coverslips, glass covers, slide covers
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.16
Prodotti consigliati
Materiali
borosilicate glass
rectangular
Confezionamento
pack of 100 ea
case of 1000 ea
Produttore/marchio commerciale
Corning 2980-225
Largh. × lungh.
22 mm × 50 mm
Tipo di binding
non-treated surface
Caratteristiche e vantaggi
- Corning® cover glasses are made from optically clear borosilicate glass.
- Chemically resistant cover glasses are not affected by weathering or surface attachment.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
CLS2980225-1000EA | 4061831696868 |
CLS2980225-100EA | 4061831696875 |