BR470045
BRAND® cover glasses for microscope slides
L × W 18 mm × 18 mm, square
Sinonimo/i:
Cover glasses, Microscopy
About This Item
Codice UNSPSC:
41122600
NACRES:
MA.01
Prodotti consigliati
Materiali
borosilicate glass
square
Produttore/marchio commerciale
BRAND 470045
Lungh. × largh.
18 mm × 18 mm
Spessore
0.13-0.17 mm , No. 1
Categorie correlate
Descrizione generale
Clear borosilicate glass, hydrolytic class 1, with excellent chemical resistance. Thickness No. 1 (0.13 to 0.17 mm). Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Full automatic production guarantees "ready-for-use" quality - clean, dust and grease free, distortion-free, flatness within ± 3 µm.
Caratteristiche e vantaggi
- Ready-for-use
- Made of hydrolytic class 1 glass with great chemical resistance
- Clean: dust and grease free
- Distortion-free
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)