CLS285525
Corning® cover glasses
square, No. 2, W × L 25 mm × 25 mm
Sinonimo/i:
coverslips, glass covers, slide covers
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.16
Prodotti consigliati
Materiali
borosilicate glass
square
Confezionamento
pack of 100 ea
case of 1000 ea
Produttore/marchio commerciale
Corning 2855-25
Largh. × lungh.
25 mm × 25 mm
Tipo di binding
non-treated surface
Categorie correlate
Caratteristiche e vantaggi
- Corning® cover glasses are made from optically clear borosilicate glass.
- Chemically resistant cover glasses are not affected by weathering or surface attachment.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
CLS285525-1000EA | 4061833882221 |
CLS285525-100EA | 4061833882238 |