CLS11656
Corning® closed system bottle
capacity 250 mL, polystyrene, male MPC connector, sterile
Sinonimo/i:
storage vessel
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
clear bottle
polystyrene
round bottle
wide-mouth bottle
Sterilità
sterile
Caratteristiche
cap
male MPC connector
Confezionamento
pkg of 6 ea
Produttore/marchio commerciale
Corning 11656
Capacità
250 mL
Categorie correlate
Descrizione generale
Corning polystyrene Storage Bottles are now available as closed system solutions. They are sterile and arrive individually double-bagged and preassembled.
The preassembled transfer caps have two ports.
One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer Lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
The preassembled transfer caps have two ports.
One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer Lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
- Made from optically clear polystyrene
- Sterilized to SAL 10-6
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)