Key Documents
BAF369441000
Bel-Art® Earth Friendly Spoon
Long Handle, capacity 1 tsp
Sinonimo/i:
sampling spoon
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
Materiali
(PLA (polylactic acid) resin )
natural
Confezionamento
package of 10
Produttore/marchio commerciale
Bel-Art F36944-1000
Lunghezza
7 in.
Capacità
1 tsp
Descrizione generale
Lessen environmental impact with these PLA (polylactic acid) resin sampling spoons. The production of 100% plant-based PLA contributes less greenhouse gasses and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill. Use a separate spoon for each material sampled to avoid cross contamination.
Caratteristiche e vantaggi
Green alternative to petroleum-based sampling tools; helps meet sustainability goals
Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
Altre note
Long handle, 17.8cm (7”), helps prevent personal contact with sampled material when reaching into bottles, jars and other confined areas
Rim of each bowl has a flat edge to assist in collecting and measuring small amounts with accuracy
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP