Key Documents
AXYRESSW1LP
AxyVoir reservoir
1 bottom, low profile, single well, 25 ea of
Sinonimo/i:
storage reservoir
About This Item
Codice UNSPSC:
41121522
NACRES:
NB.22
Produttore/marchio commerciale
Axygen Inc RES-SW1-LP
Corning
Confezionamento
25 ea of