Key Documents
AXYPCR48C
Corning® Axygen® 48 Well PCR Microplate
size 48 wells, conical bottom clear polypropylene wells, pkg of 5x10plates/cs
Sinonimo/i:
Corning® Axygen® PCR Microplate
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali
conical bottom clear polypropylene wells
Sterilità
non-sterile
Caratteristiche
lid: no
skirt: no
Produttore/marchio commerciale
Axygen Inc PCR-48-C
Corning
Confezionamento
pkg of 5x10plates/cs
Volume massimo
200 μL
Misura
48 wells
Volume d’esercizio
200 μL
Compatibilità
suitable for PCR
Tipo di binding
non-treated surface
Note legali
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.