Documenti fondamentali
AXYAM96PCRRD
Sealing mat
for PCR microplates, size 96 wells, pkg of 5x10mats/cs
Sinonimo/i:
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate
About This Item
Codice UNSPSC:
41122410
NACRES:
NB.22
Materiali
silicone (research-grade)
Sterilità
non-sterile
Produttore/marchio commerciale
Axygen Inc AM-96-PCR-RD
Corning
Confezionamento
pkg of 5x10mats/cs
Misura
96 wells
Descrizione generale
  • Axygen® AxyMats® 96 Round Well Sealing Mats are ideal for a wide range of storage and PCR applications.
  • Autoclavable mats are made of research-grade silicone.
  • Offers ultimate protection for storage applications involving more aggressive solvents.
  • The mats provide a tight seal that prevents evaporation of the samples.
  • Reusable - following a bleach wash and ethanol rinse protocol.
Note legali
AxyMats is a registered trademark of Corning, Inc.
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)