Documenti fondamentali
ZSSV008T1
Milli-Q
Sanitary Sampling Valve
for use with AFS®
About This Item
Codice UNSPSC:
41104200
NACRES:
JA.13
Confezionamento
pkg of 1 unit
Produttore/marchio commerciale
AFS®
Compatibilità
for use with AFS®
Descrizione generale
The sanitary sampling valve allows safe and reliable water sampling for microbiological assays. The valve remains sterile between 2 samplings. For use with AFS® Essential water purification systems.
Caratteristiche e vantaggi
  • No need to decontaminate the sampling valve by heating with flame.
  • Easy and safe sampling.
  • Sampling is done in the center of water flux, allowing reliable water sampling.
Note legali
AFS is a registered trademark of Merck KGaA, Darmstadt, Germany