Z760331
Ohaus® MB-23 and MB-25 moisture analyzers
Model MB25, AC/DC input 230 V AC, European 2-pin plug
Sinonimo/i:
moisture meters
About This Item
Codice UNSPSC:
41111502
NACRES:
NB.71
Prodotti consigliati
Produttore/marchio commerciale
Ohaus® 80252472 EU
Capacità di pesata
110 g
Parametri
50-160 °C temp. limit
W × H × depth
17 cm × 13 cm × 28 cm
Diam. piattaforma
90 mm
Descrizione generale
The MB25 has a halogen lamp for fast, even heating.
The Moisture analyzers combine high quality construction and easy setup. The LCD display is backlit, and it has a standard RS232 port.
- easy to clean design
- heating chamber is low maintenance
- compact footprint
Note legali
Ohaus is a registered trademark of Ohaus Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z760331-1EA | 4061834790556 |