Documenti fondamentali
Z742413
High pressure adapter for Q-Tube
for 12 mL Q-Tube, max. pressure 180 psi
About This Item
Codice UNSPSC:
51111800
NACRES:
NB.51
Materiali
aluminum
Produttore/marchio commerciale
Q Labtech 100-TA0180-120
Parametri
-200 psi pressure (max)
180 psi max. pressure
Descrizione generale
Pressure Adapter-12 mL-180 psi is designed to fit on 12 mL systems. This blue anodized aluminum adapter can hold up to 180 psi internal pressure. This higher threshold is ideal for hydrogenation and reactions involving ammonia hydroxide, methylamine, dimethylamine, formaldehyde, acetaldehyde, and hydrogen chloride.
Note legali
Q-Tube is a trademark of Q Labtech LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)