Z692964
Cargo interlocking manual pipette rack
magnets connect the racks securely and safely to metal surfaces
Sinonimo/i:
laboratory racks, pipette rack, serological pipette rack, storage racks
About This Item
Codice UNSPSC:
41122801
NACRES:
NB.35
Prodotti consigliati
Caratteristiche
Holds 0.1 to 50 mL pipettes
Confezionamento
pkg of 3 ea
Produttore/marchio commerciale
Heathrow Scientific HS20615M
Categorie correlate
Descrizione generale
Ideal for your serological pipettes!
The Cargo manual pipette rack fulfills your laboratory needs. Click together horizontally with side connections. Securely lock in place with top connections. Magnets on both sides of units connect securely and safely to metal surface such as file cabinets, fume hoods, refrigerators, and freezers.
Angled shelves store your 0.1 to 50 mL pipettes within arm′s reach for use. Protective lid encloses the unit to prevent dust build up. The lid also provides an additional shelf for extra space when connected horizontally. Varying shades of blue brings diversity to the lab.
The Cargo manual pipette rack fulfills your laboratory needs. Click together horizontally with side connections. Securely lock in place with top connections. Magnets on both sides of units connect securely and safely to metal surface such as file cabinets, fume hoods, refrigerators, and freezers.
Angled shelves store your 0.1 to 50 mL pipettes within arm′s reach for use. Protective lid encloses the unit to prevent dust build up. The lid also provides an additional shelf for extra space when connected horizontally. Varying shades of blue brings diversity to the lab.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z692964-3EA | 4061832769851 |