Z691038
Quickfit® flat flange lids for reaction flasks
flange bore 75 mm, center joint: ST/NS 34/35
Sinonimo/i:
DA5/75
About This Item
Prodotti consigliati
Materiali
borosilicate glass
Giunto
center joint: ST/NS34/35
Sterilità
non-sterile
Confezionamento
case of 1
Produttore/marchio commerciale
Quickfit DA5/75
Disponibilità
not available in USA and Canada
Diam. int. flangia
75 mm
Descrizione generale
Single and multi-socket lids for reaction flasks for simple extraction, reflux or for the introduction of a variety of stirrers, dropping funnels, temperature probes etc.
Use clip to secure lid to flasks, purchase separately.
Use clip to secure lid to flasks, purchase separately.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)