Z303887
Quickfit® flat flange lids for reaction flasks
center joint: ST/NS 29/32
Sinonimo/i:
DA4/100
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
borosilicate glass
Giunto
center joint: ST/NS29/32
Sterilità
non-sterile
Confezionamento
pack of 1
Produttore/marchio commerciale
Quickfit DA 4/100
Disponibilità
not available in USA and Canada
Diam. int. flangia
100 mm
Descrizione generale
Single and multi-socket lids for reaction flasks for simple extraction, reflux or for the introduction of a variety of stirrers, dropping funnels, temperature probes etc.
Use clip to secure lid to flasks, purchase separately.
Use clip to secure lid to flasks, purchase separately.
Caratteristiche e vantaggi
• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Also known as reflux or bulb condensers• The inner bulb sections provide an increased cooling surface• Ideal for reflux and Soxhlet extraction applications• Supplied with detachable plastic screw thread connectors which allow 9mm bore flexible tubing to be fitted easily and safely
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)