Z680087
MyBlock™ mini dry bath vehicle power adapter
AC/DC input 12 V AC
Sinonimo/i:
block dry, dry baths, dry blocks, heat blocks
About This Item
Codice UNSPSC:
41102404
NACRES:
NB.76
Prodotti consigliati
Produttore/marchio commerciale
Benchmark Scientific BSH100-A12
Descrizione generale
To aid in that portability an optional 12 V adapter is available that will let you power up the MyBlock mini dry bath/block heater in a car, RV, Bus, even a boat. You can also use 12 V field batteries with the standard cigarette lighter adapter as a power source.
Note legali
MyBlock is a trademark of Benchmark Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z680087-1EA | 4061833584248 |