Z677442
Camatril™ Velours nitrile flock lined gloves
size M
Sinonimo/i:
reusable glove
About This Item
Prodotti consigliati
Materiali
green gloves
green nitrile
Misura
M
Sterilità
non-sterile
Caratteristiche
CE compliant
disposable
latex free
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Confezionamento
package of 100 ea
Produttore/marchio commerciale
KCL 730 - 08
KCL 730
Lungh. × spessore
300 mm × 0.4 mm
Descrizione generale
Good flexibility, anatomical design and excellent fit ensure a good sense of touch. The flock lining offers additional wearing comfort.
- Ergonomic fit ensures good sensitivity
- Roughened palm for excellent grip when wet
- High mechanical resistance
- Cotton flock lining improves comfort
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- CE compliant
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z677442-100EA | 4061833069615 |