Z674265
Duran® GLS 80 caps and accessories
GL 80 blue PP quick release cap
Sinonimo/i:
1112716
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Prodotti consigliati
Produttore/marchio commerciale
DURAN® 1112716
Categorie correlate
Descrizione generale
Permits opening and closing of the DURAN® GLS 80® bottle with only a three-quarter turn. A matching PP pouring ring is also available, permitting clean, drip-free use.
Caratteristiche e vantaggi
- Triple thread permits user friendly opening and closing with only a three-quarter turn
- Cap offer a linerless, integral lip seal
- Pouring ring for drip-free pouring and clean, safe working
Note legali
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z674265-10EA | 4061832793191 |
Z674265-1EA |