Z562904
Wilmad® external reference system
complete set
Sinonimo/i:
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
Codice UNSPSC:
41121705
NACRES:
SB.32
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Wilmad 516-CC-5
Parametri
<600 MHz frequency
Diametro
5 mm
Categorie correlate
Descrizione generale
Perfect for isolating your reference solution from your sample, this double layered tube provides extra protection for toxic samples in NMR spectroscopy as well.
Each insert fits with precision into the outer tube.The flat bottom forces the reference solution upwards between the inner and outer tubes.
Simple to use, just place your sample in the insert and add a few drops of reference solution to the bottom of the outer tube. When pushing the inner tube downward, the reference solution is forced upwards and forms a thin film, about 50 μL thick between the tubes.
Each insert fits with precision into the outer tube.The flat bottom forces the reference solution upwards between the inner and outer tubes.
Simple to use, just place your sample in the insert and add a few drops of reference solution to the bottom of the outer tube. When pushing the inner tube downward, the reference solution is forced upwards and forms a thin film, about 50 μL thick between the tubes.
Note legali
Wilmad is a registered trademark of SP Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)