Documenti fondamentali
Z548987
Sigma-Aldrich
Aldrich® splash protector
female joint: ST/NS 29/32, male joint: ST/NS 24/40
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
female joint: ST/NS29/32
male joint: ST/NS24/40
Categorie correlate
Descrizione generale
Effectively prevents upward movement of foaming or bumping liquids in rotary evaporator traps. Unique design has two baffles with horizontally opposing holes that may also be used with other distillation techniques.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z548987-1EA4061832117638