Z545481
Molymod® Individual organic model set
DNA (includes stand, 11cm H)
About This Item
Codice UNSPSC:
60104506
NACRES:
NB.17
Prodotti consigliati
Descrizione
base-pairs 2
Produttore/marchio commerciale
Molymod
Descrizione generale
The mushroom-shaped hydrogen Atomlink™ components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Note legali
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z545481-1SET | 4061833078389 |