Z530301
Aldrich® straight vacuum-distilling adapter
joint: ST/NS 29/32
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Prodotti consigliati
Giunto
joint: ST/NS29/32
Categorie correlate
Descrizione generale
With side hose connection and drip-tip. Improves efficiency of evaporation of solvents under reduced pressure when placed between the condenser of a rotary evaporator and a chilled receiving flask. Plug the condenser vacuum take-off exit opposite the receiving flask to eliminate path competition and direct all distillate to the chilled collection flask. SJ 35/25 adapter fits standard Büchi rotary evaporator glassware.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z530301-1EA | 4061832112138 |