Z508497
Ace flat-bottom reaction flasks
capacity 1,000 mL, w/ O-ring groove
Sinonimo/i:
bottom flask, flat flask, reaction flasks
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
flat bottom
glass
single-neck
Sterilità
non-sterile
Caratteristiche
closure type Lid
Confezionamento
pack of 1
Produttore/marchio commerciale
Ace Glass 6511-53
Capacità
1,000 mL
Categorie correlate
Descrizione generale
Cylindrical heavy-wall reaction flask with a flat bottom rounded into side of wall, and extra-thick flat flange. No constriction at top facilitates introduction or removal of material and allows easy cleaning. Optional O-ring groove allows use of CAPFE O-rings rather than gaskets. Flask is supplied with one CAPFE O-ring. Order reaction flask heads and clamps separately.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z508497-1EA | 4061832349930 |