Z408476
Scienceware® Vibrasorb® vibration damping mount
size 13 3/4 in. × 13 3/4 in.
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.71
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art H18386-0001
Misura
13 3/4 in. × 13 3/4 in.
Peso
42 lb
Descrizione generale
Isolates equipment from disturbing vibrations down to 9 Hz. Molded of dense, 2 ½ in. thick, black and white terrazzo with a polished surface that resists scratches and chemicals. Supported by four Vibro-Absorber cushions and neoprene feet. Overall height is 3 in. Supports 16 kg.
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vibrasorb is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z408476-1EA | 4061832264080 |