Documenti fondamentali
Z217050
Sigma-Aldrich
Scienceware® Vibrasorb® vibration damping mount
size 18 in. × 22 in.
Sinonimo/i:
Vibration dampening mount, Vibration isolator
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.71
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art H18386-0000
Misura
18 in. × 22 in.
Peso
87 lb
Descrizione generale
Isolates equipment from disturbing vibrations down to 9 Hz. Molded of dense, 2 ½ in. thick, black and white terrazzo with a polished surface that resists scratches and chemicals. Supported by four Vibro-Absorber cushions and neoprene feet. Overall height is 3 in. Supports 16 kg.
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vibrasorb is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)