Documenti fondamentali
Z284270
Spinbar® Pyrex® magnetic stir bars
size 1 1/2 in. × 3/8 in.
Sinonimo/i:
Stirbar, Stirring Bar, glass Stir Bar
About This Item
Codice UNSPSC:
41103806
NACRES:
NB.72
Materiali
cylindrical
round glass coated
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F37101-0112
Lunghezza
38 mm
Diametro
9.5 mm
Misura
1 1/2 in. × 3/8 in.
Descrizione generale
  • Spinbar® Pyrex® magnetic stir bars are manufactured from Type I borosilicate glass.
  • Ideal for use in applications requiring high temperature up to 274°C (525°F).
  • The smooth Pyrex®glass coating ensures easy cleaning and offers “zero absorption” thus preventing cross-contamination of samples.
  • Autoclavable.
Note legali
PYREX is a registered trademark of Corning, Inc.
Spinbar is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)