Z280003
Norprene® A-60-G laboratory tubing
I.D. × O.D. 3/8 in. × 5/8 in.
Sinonimo/i:
Saint Gobain AFL00029, Tygon A-60-G
About This Item
Codice UNSPSC:
41105108
NACRES:
NB.35
Prodotti consigliati
Confezionamento
pkg of 50 feet
Produttore/marchio commerciale
Tygon
Diam. int. × diam. est.
3/8 in. × 5/8 in.
Misura parete
1/8 in.
Categorie correlate
Descrizione generale
This chemical pump tubing is a thermoplastic elastomer tubing with chemical compatibilities approaching fluoroelastomers and silicone tubing. Black, rubber-like tubing is heat sealable. Max. temp.: 275°F (135°C).
- Chemical resistance to oxidizing agents, acids, and alkalies
- Resistance to flex-fatigue, will not weaken or crack
Note legali
Norprene is a registered trademark of Saint-Gobain Abrasives, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z280003-1PAK | 4061833578636 |