Documenti fondamentali
Z279927
Sigma-Aldrich
Norprene® A-60-G laboratory tubing
I.D. × O.D. 3/16 in. × 3/8 in.
Sinonimo/i:
Saint Gobain AFL00013, Tygon A-60-G
About This Item
Codice UNSPSC:
41105108
NACRES:
NB.35
Confezionamento
pkg of 50 feet
Produttore/marchio commerciale
Tygon
Diam. int. × diam. est.
3/16 in. × 3/8 in.
Misura parete
3/32 in.
Descrizione generale
This chemical pump tubing is a thermoplastic elastomer tubing with chemical compatibilities approaching fluoroelastomers and silicone tubing. Black, rubber-like tubing is heat sealable. Max. temp.: 275°F (135°C).
  • Chemical resistance to oxidizing agents, acids, and alkalies
  • Resistance to flex-fatigue, will not weaken or crack
Note legali
Norprene is a registered trademark of Saint-Gobain Abrasives, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)