Z275344
Cannon-Fenske viscometer tube
size 300
About This Item
Codice UNSPSC:
41111700
NACRES:
PB.36
Prodotti consigliati
Misura
300
Descrizione
constant ~ 0.25 cSt/s
Produttore/marchio commerciale
Ace Glass 798845
Campo di misura della viscosità
50-250 cSt
Categorie correlate
Descrizione generale
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z275344-1EA | 4061832349220 |