Z256803
Ace Allihn condenser with Ace-Thred™ hose connections
joint: ST/NS 24/40, jacket L 300 mm
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Giunto
joint: ST/NS24/40
Produttore/marchio commerciale
Ace Glass 5946118
Lunghezza camicia
300 mm
Categorie correlate
Descrizione generale
Fitted with #7 Ace-Thred hose connections for 0.25 in. I.D. tubing. Features threaded inlet/outlet connections that incorporate UHDPE (ultra-high-density PE) connectors and nylon bushings to make easy connect/disconnect of coolant tubing. The bushing compresses the connector and a silicone O-ring for a leaktight seal. Approximately one bulb per 50mm.
Note legali
Ace-Thred is a trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z256803-1EA | 4061832362991 |