Documenti fondamentali
BR152505
BRAND® tubing adapter
high-density polyethylene, for tubing i.d., 8-10 to 11-14 mm
Sinonimo/i:
Tubing, Tubing adapters
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Materiali
high-density polyethylene
Caratteristiche
for tubing i.d. 8-10 to 11-14 mm
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
BRAND 152505
Lunghezza
66 mm
Categorie correlate
Descrizione generale
The BRAND® tubing adapter is designed to provide a secure connection. Its translucent construction allows for easy visibility of fluid flow, facilitating monitoring during use.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)