Z218901
Ace giant extraction thimble
I.D. × L 68 mm × 250 mm
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Produttore/marchio commerciale
Ace Glass 681204
Diam. int. × lungh.
68 mm × 250 mm
Categorie correlate
Descrizione generale
For use with Ace giant Soxhlet extractors. Thimbles are readily permeable to the flow of ether and other organic solvents. Made of cellulose.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)