Documenti fondamentali
Z180629
Ace European-style three-neck flask
capacity 250 mL
Sinonimo/i:
bottom flask, flask round, round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
flask
three-neck
Giunto
center joint ST/NS29/42
side joint ST/NS24/40
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper Joint
Confezionamento
pack of 1
Produttore/marchio commerciale
Ace Glass 6961-40
Capacità
250 mL
Descrizione generale
Has tapered walls with shallow bottom for more efficient magnetic or mechanical stirring with half-moon shaped paddle stirrer. 7 mm O.D. threaded thermometer port with nylon bushing and FETFE® O-ring.
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)