Z158399
Ace jacketed short-path distillation apparatus
Still body, with Vigreux indents
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
Prodotti consigliati
Produttore/marchio commerciale
Ace Glass 931760
Categorie correlate
Descrizione generale
For small quantities. Jacketed still body, with 10/18 thermometer joint, one-piece 14/20 cow receiver, and 10 mL distillation flask with 14/20 top opening. Order thermometer adapter Z125350 and thermometer separately.
Replacement parts:
Z158364, Jacketed still body, plain
Z158372, Jacketed still body, with Vigreux indents
Replacement parts:
Z158364, Jacketed still body, plain
Z158372, Jacketed still body, with Vigreux indents
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z158399-1EA | 4061832347981 |