Key Documents
Z118923
Safety bulb®
pkg of 1 ea
Sinonimo/i:
pipette bulb, pipette filler
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Materiali
rubber
silicone
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art H37888-0000
Descrizione generale
Large capacity, for all pipette sizes. Tapered silicone seal prevents loss of suction. May be opened for cleaning; autoclavable. Elastic cord provided for hanging on reagent bottles.
Note legali
Safety bulb is a registered trademark of SP Scienceware, a division of SP
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Pipette bulb volume 1 mL
Sigma-Aldrich
Z509035
Pipette bulb
Pipette bulb capacity 5 mL
Z186678
Pipette bulb
Pipette bulb capacity 2 mL
Sigma-Aldrich
Z136042
Pipette bulb
RF1000™ pipette filler a lightweight manual pipette filler uniquely designed for one-handed operation
Z675997
RF1000 pipette filler
Pipette fillers red silicone bulb type
Z239089
Pipette fillers
Pipette bulb volume 3 mL
Sigma-Aldrich
Z509043
Pipette bulb
Squeeze-bulb pump internal flapper valve prevents reverse flow, pkg of 1 ea
Z200336
Squeeze-bulb pump
BRAND® pipette fillers Standard, for pipettes up to 10 mL
Z333980
BRAND® pipette fillers
Pipette fillers red natural rubber bulb type
Z136468
Pipette fillers