Z118311
Aldrich® large-capacity cold trap
Replacement bulb, for Z103101, joint: ST/NS 55/50
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
Prodotti consigliati
Giunto
joint: ST/NS55/50
Capacità bulbo
500 mL
Categorie correlate
Descrizione generale
With removable cold-fingers and a choice of 55/50 joint or 50 mm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z118311-1EA | 4061832131313 |