Z115231
Scienceware® piezoelectric lighter
safe, useful tool for igniting Bunsen burners, gas stoves, propane, and oxyacetylene torches
Sinonimo/i:
piezo ignitor, piezo lighter, piezoelectric ignitor
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
Prodotti consigliati
Produttore/marchio commerciale
Bel-Art
Misura
1 in. × 1 1/2 in. × 8 in.
Descrizione generale
A safe, useful tool for igniting Bunsen burners, gas stoves, propane, and oxyacetylene torches. Produces a burst of 6 to 8 electric sparks at 20 kV without batteries, fuels, or flints. Plastic handle and a mounting bracket for convenient storage or hanging.
Altre note
When using the product, the end of the barrel should be inserted in the middle of the actual gas flow, not off to the side. The gas needs to flow through the holes in the end of the barrel. Please note that the spark isn′t very large.
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)