Documenti fondamentali
Z102350
Perfektum® one-way compression-nut stopcock
Female Luer to male Luer lock, not unidirectional
Sinonimo/i:
Compression-Nut Stopcock, One-Way Compression Stopcock, Stopcock for Compression
About This Item
Codice UNSPSC:
41122000
NACRES:
NB.03
Produttore/marchio commerciale
Cadence Science 6021
Descrizione generale
Water tested at 80 psi.
Note legali
Perfektum is a registered trademark of Cadence Science
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 8
1 of 8
3-way stopcock female Luer lock to male Luer lock, left side, female Luer lock
CAD6017
3-way stopcock
1-way stopcock male Luer lock to 8 mm hose end
CAD6030
1-way stopcock
PTFE Body Two-Way Valve for use with Hamilton PTFE Luer Lock (TLL) and other Luer lock syringes.
20926
PTFE Body Two-Way Valve
3-way stopcock female Luer lock to male Luer lock, right side, female Luer lock, compression-nut
CAD6011
3-way stopcock
SGE Syringe Valves Push button with Luer Lock, for use with SGE 5mL-100mL capacity syringe, volume 5-2000 mL
509353
SGE Syringe Valves
3-way stopcock female Luer lock to male Luer lock, left side, female Luer lock, stainless steel
CAD6017SS
3-way stopcock
Global Trade Item Number
SKUGTIN
Z102350-1EA4061832529745