Documenti fondamentali
WIMWG1209J2
Multi-tube jet washer/dryer, 5-position, for 3-5 mm NMR tubes
joint: ST/NS 29/32
Sinonimo/i:
cleaning tube washers, tube washers
About This Item
Codice UNSPSC:
47131800
NACRES:
SB.32
Giunto
joint: ST/NS29/32
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Wilmad WG-1209-J2
Descrizione generale
This Wilmad Multi-Tube Jet Solvent Washer/Dryer is recommended for research labs that routinely clean NMR and EPR tubes; a single unit can accommodate up to 5 tubes (3, 4 and 5 mm OD) at once. When an inverted tube is inserted onto the solvent transfer tubing and the open end is immersed under wash solvent in the solvent cup, a reliable vacuum-tight seal will be formed and generate solvent flow under vacuum. After solvent is fully consumed, air flow will follow to turn the unit into a dryer.
Features:
  • 5 PTFE coated stainless steel solvent transfer tubes fit 3, 4 and 5 mm NMR/EPR tubes
  • PTFE solvent cup and tubing make this unit fully resistant to common organic solvents
  • Flanged reservoir connection eliminates joint freeze
  • Greaseless joint between the solvent cup and glass reservoir eliminates possibility of contamination
  • Complete disassembly without tools for easy cleaning
  • Hands free during washing/drying cycle
  • Calibrated length mark for 4 in., 7 in., 8 in. and 9 in. tube
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2