Z553336
Washer for NMR tubes with J. Young valve
top joint: ST/NS 19/26, bottom joint: ST/NS 24/40
About This Item
Codice UNSPSC:
47131800
NACRES:
SB.32
Prodotti consigliati
Giunto
bottom joint: ST/NS24/40
top joint: ST/NS19/26
Descrizione generale
The inverted 5 mm NMR tube is sealed into the washer with a ground-glass stopper. Applied vacuum sprays solvent inside and against the closed end of tube for thorough cleaning.
Fits flask with standard taper 24/40 joint.
Fits flask with standard taper 24/40 joint.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z553336-1EA | 4061833305461 |