Documenti fondamentali
WHA77041001
Whatman® microplate lid
Clear polystyrene universal lid
Sinonimo/i:
Z746932, multiwell plate lid
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.24
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
Whatman 7704-1001
Whatman Article No. 28420899 (US reference)
Descrizione generale
Lids are suitable for using as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)