TEM-LC400CUUL
Ultrathin Lacey Carbon Supported Copper Grid
size 400 mesh, box of 25
Sinonimo/i:
LACEY CARBON FILM CU #400 ULTRA THIN 25/BX
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Prodotti consigliati
Confezionamento
box of 25
Misura
400 mesh
Categorie correlate
Descrizione generale
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Copper Grid, 400 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)