TEM-LC200NIUL
Ultrathin Lacey Carbon Supported Nickel Grid
size 200 mesh, box of 25
Sinonimo/i:
LACEY CARBON FILM - NI 200 MESH ULTRA THIN 25/BX
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Prodotti consigliati
Confezionamento
box of 25
Misura
200 mesh
Descrizione generale
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Nickel Grid, 200 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
TEM-LC200NIUL | 4061842455591 |