Documenti fondamentali
SYNC562280
Synthware coil-style reflux condenser with removable hose connections
joint: ST/NS 14/20, hose size 8 mm
Sinonimo/i:
Synthware C562280, coil-type condenser
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C562280
Dimens. tubo
8 mm
Descrizione generale
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)