Documenti fondamentali
SYNA842415
Synthware vacuum filtration adapter
bottom joint: ST/NS 24/40, Stopper Size: #3, joint: ST/NS 24/40, stopper size #3
Sinonimo/i:
Synthware A842415, Glass adapter vacuum filtration
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Giunto
bottom joint: ST/NS24/40
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
A842415
Synthware A842415
Descrizione generale
Used with plain stem Buchner funnels and pluro-stopper. Hose connection has an O.D of 10 mm at the largest serration. Order pluro-stoppers below separately.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)