SYNA502440
Synthware™ gas inlet adapter
joint: ST/NS 24/40, O.D. × L 153 mm
Sinonimo/i:
Synthware A502440, gas inlet adapter, extended drip tube
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Prodotti consigliati
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A502440
D.E. × lungh.
153 mm
Categorie correlate
Descrizione generale
Inner joint with a sealed on lower tube extending 6” below the grind, having the same O.D. tubing on both ends of the grind.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)