Documenti fondamentali
CLS5340250
Pyrex® heavy wall graduated filtering flask, with tubulation
10 mm O.D. hose-connection, capacity 250 mL
Sinonimo/i:
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 5340
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
flat bottom flask
narrow-mouth
Giunto
joint stopper6
Sterilità
non-sterile
Caratteristiche
closure type Rubber stopper
Confezionamento
case of 18
pack of 6
Produttore/marchio commerciale
Corning 5340-250
Campo di misura del volume
75-250 mL
Capacità
250 mL
Descrizione generale
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)