Documenti fondamentali
CLS4450250
Pyrex® baffled Erlenmeyer flask
capacity 250 mL
Sinonimo/i:
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, laboratory flasks, Corning® 4450
About This Item
Codice UNSPSC:
41122104
eCl@ss:
32040501
Materiali
borosilicate glass
flat bottom flask
narrow-mouth
narrow-mouth neck
Sterilità
non-sterile
Caratteristiche
cap: no
closure type Rubber stopper
Confezionamento
case of 6
Produttore/marchio commerciale
Corning 4450-250
Capacità
250 mL
Descrizione generale
  • Pyrex® baffled Erlenmeyer flasks feature a heavy tooled rim for maximum durability.
  • Baffle-designed Delong flasks promote optimal and consistent solution agitation when used with flask shakers.
  • Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.
  • Includes a marking spot for convenience.
Altre note
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)