CAD9917
Animal feeding needle, sterile, disposable, malleable 304 SS
size 18G, L × diam. 3.1 in. × 2.3 mm , ball
Sinonimo/i:
304 SS Feeding Needle, Malleable Feeding Needle
About This Item
Codice UNSPSC:
42142609
NACRES:
NB.03
Prodotti consigliati
Misura
18G
Confezionamento
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diam.
3.1 in. × 2.3 mm , ball
Categorie correlate
Descrizione generale
- Designed to enable the researcher to shape the needle to specific needs
- Needles come straight and are easily bent to any desired shape
- Made of type 304 stainless steel tubing
- Plastic Luer hubs
- Silicone tips for smoothest possible insertion
- Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)