Documenti fondamentali
BR95111
BRAND® special pegs for draining rack
Sinonimo/i:
Accessories, Draining rack, Special pegs
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.35
Materiali
polystyrene
Confezionamento
pack of 1 set
pack of 11 ea
Produttore/marchio commerciale
BRAND 95111
Lungh. × D.E.
100 mm × 6 mm
Descrizione generale
The BRAND® special pegs for the draining rack are crafted from PS and are specifically designed to accommodate narrow-mouthed or small vessels, such as test tubes, small volumetric flasks, and graduated cylinders. These pegs provide a secure and stable hold, ensuring that your vessels drain effectively while minimizing the risk of spills or breakage. Their thoughtful design allows for efficient use of space on the draining rack, making them an essential accessory for laboratory environments.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)